APO455833258 Apollo Scientific Ltd

Ethyl (4-bromophenoxy)acetate

  • MDL number MFCD00462746
  • Cas Number : 6964-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4-Bromophenoxy)acetic acid ethyl ester, Ethyl 2-(4-bromophenoxy)ethanoate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36605

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36605
pictograms GHS05;GHS07
hstatements H302+H312+H332;H318
Availability details for
Quantity
  • Availability
  • Add to Cart