APO455833262 Apollo Scientific Ltd

5-Bromo-2-(piperazin-1-yl)pyridine

  • MDL number MFCD04115056
  • Cas Number : 73406-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36612

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36612
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart