APO455833266 Apollo Scientific Ltd

8-Chloroisoquinoline

  • MDL number MFCD07644622
  • Cas Number : 34784-07-1
  • Molecular Weight : 163.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Chloro-2-azanaphthalene
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-chloroisoquinoline

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/8-chloroisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart