APO455833269 Apollo Scientific Ltd

3-Chloro-2-methoxypyridine

  • MDL number MFCD03095227
  • Cas Number : 13472-84-9
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2-methoxypyridine
density 1.25
Inchi Code 1S/C6H6ClNO/c1-9-6-5(7)3-2-4-8-6/h2-4H,1H3
Inchi Key MSICGKUDCSUMJZ-UHFFFAOYSA-N

Description

Purity 98%
Density 1.25
Flash Point 50-53/3mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-methoxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-methoxypyridine
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart