APO455833271 Apollo Scientific Ltd

6-Methoxyisoquinoline

  • MDL number MFCD04114861
  • Cas Number : 52986-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Methoxy-2-azanaphthalene
Storage temp. Store at 2-8
Physical Form Liquid-Viscous
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 88-95/0.8mm
Product Link https://store.apolloscientific.co.uk/product/6-methoxyisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-methoxyisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart