APO455833283 Apollo Scientific Ltd

3-Chloro-4-hydroxybenzeneboronic acid

  • MDL number MFCD09258739
  • Cas Number : 182344-13-4
  • Molecular Weight : 172.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 223-228
Iupac Name 3-chloro-4-hydroxyphenylboronic acid
Inchi Code 1S/C6H6BClO3/c8-5-3-4(7(10)11)1-2-6(5)9/h1-3,9-11H
Inchi Key WWQIKFZZILXJHG-UHFFFAOYSA-N

Description

Synonym 4-Borono-2-chlorophenol
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 223-228
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-hydroxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-hydroxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart