APO455833285 Apollo Scientific Ltd

1,2-Dihydro-4-hydroxy-2-oxopyridine-3-carbonitrile

  • MDL number MFCD09258786
  • Cas Number : 5657-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-1,2-dihydro-4-hydroxy-2-oxopyridine, 1,2-Dihydro-4-hydroxy-2-oxonicotinonitrile
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36741

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36741
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart