APO455833290 Apollo Scientific Ltd

5-Bromo-2-(pyrrolidin-1-yl)pyrimidine

  • MDL number MFCD00483243
  • Cas Number : 446286-61-9
  • Molecular Weight : 228.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-(1-pyrrolidinyl)pyrimidine
Inchi Code 1S/C8H10BrN3/c9-7-5-10-8(11-6-7)12-3-1-2-4-12/h5-6H,1-4H2
Inchi Key AKQGPROZPHKDMD-UHFFFAOYSA-N

Description

Synonym 1-(5-Bromopyrimidin-2-yl)pyrrolidine, 5-Bromo-2-(pyrrolidin-1-yl)-1,3-diazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-pyrrolidin-1-ylpyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-pyrrolidin-1-ylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart