APO455833298 Apollo Scientific Ltd

2-Amino-6-bromonaphthalene

  • MDL number MFCD01026466
  • Cas Number : 7499-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-2-naphthylamine, 6-Bromonaphthalen-2-amine
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36487

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=36487
Availability details for
Quantity
  • Availability
  • Add to Cart