APO455833300 Apollo Scientific Ltd

N,N,N',N'-Tetracyclohexylsebacamide

  • MDL number MFCD08445500
  • Cas Number : 5426-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N,N',N'-Tetracyclohexyldecane-1,10-diamide
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16103

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16103
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart