APO455833319 Apollo Scientific Ltd

5-(Morpholin-4-yl)thiophene-2-carboxaldehyde

  • MDL number MFCD02710611
  • Cas Number : 24372-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-128
Synonym 2-Formyl-5-(morpholin-4-yl)thiophene
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-morpholin-4-ylthiophene-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS https://store.apolloscientific.co.uk/product/5-morpholin-4-ylthiophene-2-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart