APO455833323 Apollo Scientific Ltd

Methyl 3-(5-bromopyridin-3-yl)-3-oxopropanoate

  • MDL number MFCD00216527
  • Cas Number : 205985-97-3
  • Molecular Weight : 258.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Synonym Methyl (5-bromonicotinoyl)acetate, Methyl 3-(5-bromopyridin-3-yl)-3-oxopropionate
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-5-bromopyridin-3-yl-3-oxopropanoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-5-bromopyridin-3-yl-3-oxopropanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart