APO455833325 Apollo Scientific Ltd

1-Methyl-1H-indole-4-boronic acid, pinacol ester

  • MDL number MFCD08690266
  • Cas Number : 898289-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Synonym 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Purity 97
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-4-boronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-4-boronic-acid-pinacol-ester
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart