APO455833326 Apollo Scientific Ltd

4-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester

  • MDL number MFCD08690299
  • Cas Number : 884507-39-5
  • Molecular Weight : 287.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-50
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-pyrrolidin-1-ylmethylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-pyrrolidin-1-ylmethylbenzeneboronic-acid-pinacol-ester
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart