APO455833361 Apollo Scientific Ltd

3-(Aminomethyl)biphenyl

  • MDL number MFCD00270124
  • Cas Number : 177976-49-7
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28-30
Synonym (Biphenyl-3-yl)methylamine, 3-Phenylbenzylamine
Purity 95
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 128/1mm
Product Link https://store.apolloscientific.co.uk/product/3-aminomethylbiphenyl

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-aminomethylbiphenyl
pictograms GHS05;GHS07
hstatements H314;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart