APO455833362 Apollo Scientific Ltd

5-Amino-1-methyl-1H-pyrazole

  • MDL number MFCD00068156
  • Cas Number : 1192-21-8
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 73-76
Iupac Name 1-methyl-1H-pyrazol-5-ylamine
Inchi Code 1S/C4H7N3/c1-7-4(5)2-3-6-7/h2-3H,5H2,1H3
Inchi Key JESRNIJXVIFVOV-UHFFFAOYSA-N

Description

Synonym 1-Methyl-1H-pyrazol-5-amine, 2-Methyl-2H-pyrazol-3-amine
Purity 97%
Flash Point 95-96/0.5mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 73-76
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-methyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-methyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart