APO455833374 Apollo Scientific Ltd

Coumarin-6-carboxaldehyde

  • MDL number MFCD01941322
  • Cas Number : 51690-26-7
  • Molecular Weight : 174.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 192-194
Iupac Name 2-oxo-2H-chromene-6-carbaldehyde
Inchi Code 1S/C10H6O3/c11-6-7-1-3-9-8(5-7)2-4-10(12)13-9/h1-6H
Inchi Key RQCHYJGDSCWRSQ-UHFFFAOYSA-N

Description

Synonym 2-Oxo-2H-chromene-6-carboxaldehyde, 2-Oxo-2H-1-benzopyran-6-carboxaldehyde
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 192-194
Product Link https://store.apolloscientific.co.uk/product/coumarin-6-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/coumarin-6-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart