APO455833380 Apollo Scientific Ltd

2-Chloro-5-cyanobenzeneboronic acid

  • MDL number MFCD06656271
  • Cas Number : 936249-33-1
  • Molecular Weight : 181.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-cyanophenylboronic acid
Inchi Code 1S/C7H5BClNO2/c9-7-2-1-5(4-10)3-6(7)8(11)12/h1-3,11-12H
Inchi Key YNZXUVCJPQNYHR-UHFFFAOYSA-N

Description

Synonym 3-Borono-4-chlorobenzonitrile
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-cyanobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-cyanobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart