APO455833390 Apollo Scientific Ltd

Isopropyl toluene-4-sulphonate

  • MDL number MFCD00968877
  • Cas Number : 2307-69-9
  • Molecular Weight : 214.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
bp 20
Iupac Name isopropyl 4-methylbenzenesulfonate
density 1.142
Inchi Code 1S/C10H14O3S/c1-8(2)13-14(11,12)10-6-4-9(3)5-7-10/h4-8H,1-3H3
Inchi Key SDQCGKJCBWXRMK-UHFFFAOYSA-N

Description

Melting Point >110
Synonym Isopropyl 4-methylbenzenesulphonate, Prop-2-yl 4-methylbenzenesulphonate
Purity 98%
Density 1.142
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 20
Product Link https://store.apolloscientific.co.uk/product/isopropyl-toluene-4-sulphonate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/isopropyl-toluene-4-sulphonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart