APO455833393 Apollo Scientific Ltd

1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD00035267
  • Cas Number : 6429-04-5
  • Molecular Weight : 379.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Tetrahydropapaverine hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-34-dimethoxybenzyl-67-dimethoxy-1234-tetrahydroisoquinoline-hydrochloride

Safety Information

signalword Warning
pcodes P280;P261;P301+P312
MSDS https://store.apolloscientific.co.uk/product/1-34-dimethoxybenzyl-67-dimethoxy-1234-tetrahydroisoquinoline-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart