APO455833398 Apollo Scientific Ltd

2,6-Dimethylpyridine N-oxide

  • MDL number MFCD00039715
  • Cas Number : 1073-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Lutidine N-oxide, 2,6-Dimethylpyridinium-1-olate
Density 1.102
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 108-111/10mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32520

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32520
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart