APO455833401 Apollo Scientific Ltd

5-Bromo-2-methoxyphenol

  • MDL number MFCD00673137
  • Cas Number : 37942-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Synonym 4-Bromo-2-hydroxyanisole, 5-Bromoguaiacol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 150/20mm
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-methoxyphenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-methoxyphenol
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart