APO455833411 Apollo Scientific Ltd

4-Bromo-2-chloroiodobenzene

  • MDL number MFCD00079709
  • Cas Number : 31928-47-9
  • Molecular Weight : 317.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 33-35
Iupac Name 4-bromo-2-chloro-1-iodobenzene
Inchi Code 1S/C6H3BrClI/c7-4-1-2-6(9)5(8)3-4/h1-3H
Inchi Key OHHKQBZOURGNLR-UHFFFAOYSA-N

Description

Synonym 4-Bromo-2-chloro-1-iodobenzene
Purity 97%
Flash Point 160-162/15mm
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 33-35
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-chloroiodobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-chloroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart