APO455833427 Apollo Scientific Ltd

Diethyl 2-chloro-3-oxosuccinate

  • MDL number MFCD04114359
  • Cas Number : 34034-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Diethyl 2-chloro-3-oxobutane-1,4-dioate
Density 1.23
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 98/0.5mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32483

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32483
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart