APO455833437 Apollo Scientific Ltd

Isoquinoline-5-sulphonyl chloride hydrochloride

  • MDL number MFCD08272853
  • Cas Number : 105627-79-0
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Chlorosulphonyl)isoquinoline hydrochloride, 5-(Chlorosulphonyl)-2-azanaphthalene hydrochloride
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-5-sulphonyl-chloride-hydrochloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-5-sulphonyl-chloride-hydrochloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart