APO455833438 Apollo Scientific Ltd

6,7,8,9-Tetrahydro-5H-imidazo[1,2-a]azepine-3-sulphonyl chloride

  • MDL number MFCD08235265
  • Cas Number : 914637-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Chlorosulphonyl)-6,7,8,9-tetrahydro-5H-imidazo[1,2-a]azepine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31925

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31925
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart