APO455833441 Apollo Scientific Ltd

2,4-Dimethyl-3-hydroxypyridine

  • MDL number MFCD08235268
  • Cas Number : 27296-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-40
Synonym 2,4-Dimethylpyridin-3-ol, 3-Hydroxy-2,4-lutidine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/24-dimethyl-3-hydroxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-dimethyl-3-hydroxypyridine
pictograms GHS07
hstatements H302;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart