APO455833451 Apollo Scientific Ltd

2-Methylpiperidin-4-one, N-BOC protected

  • MDL number MFCD04035608
  • Cas Number : 190906-92-4
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-4-oxopiperidine, N-BOC protected, tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-2-methylpiperidin-4-one
Purity 97
Physical Form Solid-Fused
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methylpiperidin-4-one-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylpiperidin-4-one-n-boc-protected
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart