APO455833454 Apollo Scientific Ltd

2-Ethylpiperidin-4-one, N-BOC protected

  • MDL number MFCD07367729
  • Cas Number : 324769-07-5
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 2-ethyl-4-oxopiperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-2-ethyl-4-oxopiperidine
Purity 97
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-ethylpiperidin-4-one-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-ethylpiperidin-4-one-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart