APO455833455 Apollo Scientific Ltd

4-Methylenepiperidine, N-BOC protected

  • MDL number MFCD01632522
  • Cas Number : 159635-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 4-methylidenepiperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-methylidenepiperidine
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 75-80/1mm
Product Link https://store.apolloscientific.co.uk/product/4-methylenepiperidine-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylenepiperidine-n-boc-protected
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart