APO455833470 Apollo Scientific Ltd

[3-(4-Nitrophenyl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD00665862
  • Cas Number : 959582-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-[3-(4-Nitrophenyl)-1H-pyrazol-1-yl]ethanoic acid, 1-(Carboxymethyl)-3-(4-nitrophenyl)-1H-pyrazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15499

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15499
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart