APO455833473 Apollo Scientific Ltd

(5-Thien-2-ylisoxazol-3-yl)methylamine

  • MDL number MFCD08271879
  • Cas Number : 852180-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32015

Safety Information

signalword Danger
pcodes P310;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32015
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart