APO455833479 Apollo Scientific Ltd

3-(1,3-Dioxolan-2-yl)thiophene-2-sulphonyl chloride

  • MDL number MFCD08271969
  • Cas Number : 103011-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Synonym 2-[2-(Chlorosulphonyl)thien-3-yl]-1,3-dioxolane, 2-(Chlorosulphonyl)-3-(1,3-dioxolan-2-yl)thiophene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32054

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32054
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart