APO455833480 Apollo Scientific Ltd

2-Phenoxybenzyl alcohol

  • MDL number MFCD00017297
  • Cas Number : 13807-84-6
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Phenoxyphenyl)methanol, 2-(Hydroxymethyl)diphenyl ether
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-phenoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-phenoxybenzyl-alcohol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart