APO455833484 Apollo Scientific Ltd

4-(Aminomethyl)-2,6-dichloropyridine

  • MDL number MFCD06212867
  • Cas Number : 88579-63-9
  • Molecular Weight : 177.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73
Synonym (2,6-Dichloropyridin-4-yl)methylamine, 2,6-Dichloroisonicotinylamine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyl-26-dichloropyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyl-26-dichloropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart