APO455833486 Apollo Scientific Ltd

2-(Naphthoxy)acetic acid hydrazide

  • MDL number MFCD00156157
  • Cas Number : 24310-15-4
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-naphthyloxy)acetohydrazide
Inchi Code 1S/C12H12N2O2/c13-14-12(15)8-16-11-7-3-5-9-4-1-2-6-10(9)11/h1-7H,8,13H2,(H,14,15)
Inchi Key PLYHRLDIWQEXBD-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-naphthoxyacetic-acid-hydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-naphthoxyacetic-acid-hydrazide
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart