APO455833493 Apollo Scientific Ltd

7-Aminothieno[2,3-b]pyrazine

  • MDL number MFCD08271970
  • Cas Number : 59944-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87.5-89.5
Synonym Thieno[2,3-b]pyrazin-7-amine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32107

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32107
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart