APO455833494 Apollo Scientific Ltd

(1,3-Benzodioxol-5-yl)acetyl chloride

  • MDL number MFCD03424722
  • Cas Number : 6845-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(1,3-Benzodioxol-5-yl)ethanoyl chloride, 5-(2-Chloro-2-oxoethyl)-1,3-benzodioxole
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 110-112/0.75mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32116

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32116
pictograms GHS05;GHS07
hstatements H302;H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart