APO455833497 Apollo Scientific Ltd

3-(1H-1,2,4-Triazol-1-yl)benzaldehyde

  • MDL number MFCD08271896
  • Cas Number : 868755-54-8
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114.5-115.5
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-1h-124-triazol-1-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/3-1h-124-triazol-1-ylbenzaldehyde
pictograms GHS02;GHS07
hstatements H228;H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart