APO455833505 Apollo Scientific Ltd

5-Aminobenzo[b]furan

  • MDL number MFCD04125391
  • Cas Number : 58546-89-7
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Benzofuran-5-amine, Benzo[b]furan-5-amine
Purity 97
Storage temp. Store at -20
Physical Form Solid-Low Melt
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-aminobenzobfuran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminobenzobfuran
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart