APO455833510 Apollo Scientific Ltd

6-(Morpholin-4-yl)pyridine-2-carboxylic acid

  • MDL number MFCD03965867
  • Cas Number : 554405-17-3
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-129
Synonym 6-(Morpholin-4-yl)picolinic acid, 4-(6-Carboxypyridin-2-yl)morpholine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-morpholin-4-ylpyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-morpholin-4-ylpyridine-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart