APO455833514 Apollo Scientific Ltd

2-(2,4-Dimethylphenoxy)acetohydrazide

  • MDL number MFCD00156144
  • Cas Number : 72293-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2,4-Dimethylphenoxy)acetic acid hydrazide, 4-(2-Hydrazino-2-oxoethoxy)-m-xylene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14468

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14468
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart