APO455833516 Apollo Scientific Ltd

Methyl 4-chloro-3-nitrobenzoate

  • MDL number MFCD00016989
  • Cas Number : 14719-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-83
Synonym 2-Chloro-5-(methoxycarbonyl)nitrobenzene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-4-chloro-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-chloro-3-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart