APO455833517 Apollo Scientific Ltd

5-(3-Bromophenyl)-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD08435902
  • Cas Number : 160377-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Synonym 3-(3-Methyl-1,2,4-oxazol-5-yl)bromobenzene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32175

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32175
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart