APO455833529 Apollo Scientific Ltd

3-(4-Nitrophenyl)-1H-pyrazole

  • MDL number MFCD00665859
  • Cas Number : 20583-31-7
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-195
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-4-nitrophenyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-nitrophenyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart