APO455833537 Apollo Scientific Ltd

Tetrahydro-2H-pyran-4-carboxaldehyde

  • MDL number MFCD03425264
  • Cas Number : 50675-18-8
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Formyltetrahydro-2H-pyran, Oxane-4-carboxaldehyde
Purity 95
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tetrahydro-2h-pyran-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tetrahydro-2h-pyran-4-carboxaldehyde
pictograms GHS07
hstatements H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart