APO455833541 Apollo Scientific Ltd

Tetrahydro-2H-pyran-4-carboxamide

  • MDL number MFCD08235066
  • Cas Number : 344329-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-183
Synonym 4-Carbamoyltetrahydro-2H-pyran
Purity 98
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/tetrahydro-2h-pyran-4-carboxamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tetrahydro-2h-pyran-4-carboxamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart