APO455833544 Apollo Scientific Ltd

(2-Chloroprop-2-yl)benzene

  • MDL number MFCD00040715
  • Cas Number : 934-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1-Chloro-1-methylethyl)benzene, alpha,alpha-Dimethylbenzyl chloride
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 48-50/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14432

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14432
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart