APO455833545 Apollo Scientific Ltd

2,5-Dichlorophenylacetyl chloride

  • MDL number MFCD08543437
  • Cas Number : 203314-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32219

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32219
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart