APO455833547 Apollo Scientific Ltd

2-(Dimethylcarbamoyl)benzeneboronic acid

  • MDL number MFCD03425959
  • Cas Number : 874219-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Synonym 2-(Dimethylaminocarbonyl)benzeneboronic acid
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32224

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32224
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart